Assay Details
Target Gene Details
Entrez Gene ID: | 100039795 |
Gene Name: | immunoglobulin-like domain containing receptor 2 |
Gene Aliases: |
2810478N18Rik, 3110063L10Rik, AI852300, D1Ertd471e, Dbsm1, ENSMUSG00000040612, Ll |
Location: |
Chr.1:166254094-166316832 on Build GRCm38 |
Assay Gene Location: | Within Exon 9 |
Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
---|---|---|---|---|
Ildr2 | NM_001164528.1 | 9 | 2069 | NP_001158000.1 |
FJ024495.1 | 9 | 2015 | ||
FJ024496.1 | 8 | 1838 | ||
FJ024497.1 | 6 | 1634 | ||
FJ024498.1 | 8 | 1958 | ||
FJ024499.1 | 7 | 1691 | ||
FJ024500.1 | ||||
FJ024501.1 |
More Information
Set Membership: |
Intragenic Exonic |