Assay Details
Target Gene Details
Entrez Gene ID: | 329260 |
Gene Name: | DENN/MADD domain containing 1B |
Gene Aliases: |
4632404N19Rik, 4930467M19Rik, 6820401H01Rik, F730008N07Rik |
Location: |
Chr.1:138963446-139176043 on Build GRCm38 |
Assay Gene Location: | Within Exon 23 |
Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
---|---|---|---|---|
Dennd1b | NM_001166501.1 | 22 | 2635 | NP_001159973.1 |
NM_181347.3 | 21 | 2501 | NP_851992.1 | |
AK015524.1 | 10 | 1787 | BAB29881.1 | |
AK155727.1 | 21 | 2502 | BAE33404.1 | |
BC141388.1 | 21 | 2497 | ||
BC141389.1 | 21 | 2497 |