Assay Details
Target Gene Details
Entrez Gene ID: | 227099 |
Gene Name: | postmeiotic segregation increased 1 (S. cerevisiae) |
Gene Aliases: |
- |
Location: |
Chr.1:53189187-53297026 on Build GRCm38 |
Assay Gene Location: | Within Exon 13 |
Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
---|---|---|---|---|
Pms1 | NM_153556.1 | 13 | 2825 | NP_705784.1 |
BC028939.1 | 13 | 2825 | AAH28939.1 |
More Information
Set Membership: |
Intragenic Exonic |