Assay Details
Target Gene Details
Entrez Gene ID: | 16153 |
Gene Name: | interleukin 10 |
Gene Aliases: |
CSIF, Il-10 |
Location: |
Chr.1:131019845-131024970 on Build GRCm38 |
Assay Gene Location: | Within Exon 5 |
Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
---|---|---|---|---|
Il10 | NM_010548.2 | 5 | 1037 | NP_034678.1 |
AK152344.1 | 5 | 1038 | BAE31139.1 | |
BC120612.1 | 5 | 1012 | ||
BC137844.1 | 5 | 1012 | ||
M37897.1 | 5 | 1045 | AAA39274.1 |