Assay Details
Target Gene Details
Entrez Gene ID: | 67698 |
Gene Name: | family with sequence similarity 174, member A |
Gene Aliases: |
2310044D20Rik, Fam174, Naa6, Tmem157 |
Location: |
Chr.1:95313628-95335284 on Build GRCm38 |
Assay Gene Location: | Within Exon 3 |
Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
---|---|---|---|---|
Fam174a | NM_026321.4 | 3 | 1426 | NP_080597.2 |
AK033948.1 | 3 | 1311 | BAC28523.1 | |
AK148807.1 | 3 | 1326 | BAE28669.1 |