Assay Details
Target Gene Details
Entrez Gene ID: | 18802 |
Gene Name: | phospholipase C, delta 4 |
Gene Aliases: |
4921507K24Rik |
Location: |
Chr.1:74541972-74565977 on Build GRCm38 |
Assay Gene Location: | Within Exon 2 |
Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
---|---|---|---|---|
Plcd4 | NM_001081456.1 | NP_001074925.1 | ||
NM_148937.2 | 1 | 42 | NP_683739.2 | |
AK014836.1 | 1 | 42 | ||
AK016945.1 | 1 | 39 | BAB30513.1 | |
AK029561.1 | 1 | 37 | ||
AK039149.1 | BAC30256.1 | |||
AK140231.1 | BAE24292.1 | |||
BC066156.1 | AAH66156.1 |
More Information
Set Membership: |
Intragenic Exonic |