Assay Details
Target Gene Details
Entrez Gene ID: | 11931 |
Gene Name: | ATPase, Na+/K+ transporting, beta 1 polypeptide |
Gene Aliases: |
Atp4b, Atpb, Atpb-1, NKbeta1 |
Location: |
Chr.1:164437099-164458355 on Build GRCm38 |
Assay Gene Location: | Within Exon 6 |
Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
---|---|---|---|---|
Atp1b1 | NM_009721.6 | 6 | 2489 | NP_033851.1 |
AK005071.1 | 6 | 2136 | BAB23795.1 | |
AK140862.1 | 6 | 2136 | BAE24500.1 | |
AK160405.1 | 6 | 2135 | BAE35773.1 | |
AK161891.1 | 6 | 2140 | BAE36624.1 | |
AK162938.1 | 3 | 1478 | BAE37124.1 | |
BC027319.1 | 6 | 2128 | AAH27319.1 | |
BC094070.1 | 6 | 2127 | AAH94070.1 | |
X61433.1 | 5 | 1895 | CAA43675.1 |
Target Gene Details
Entrez Gene ID: | 171567 |
Gene Name: | NME/NM23 family member 7 |
Gene Aliases: |
D530024H21Rik, Nm23-M7, Nm23-r7 |
Location: |
Chr.1:164307662-164437300 on Build GRCm38 |
Assay Gene Location: | Within Exon 15 |
Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
---|---|---|---|---|
Nme7 | NM_001293313.1 | 14 | 1637 | NP_001280242.1 |
NM_138314.4 | 14 | 1640 | NP_612187.2 | |
AF202048.1 | 13 | 1541 | AAF20906.1 | |
AK144915.1 | 14 | 1641 | BAE26132.1 | |
AK160566.1 | 6 | 596 | ||
BC038021.1 | 15 | 1667 | ||
BC049225.1 | 13 | 1568 | AAH49225.1 | |
BU609451.1 | 1 | 102 | ||
CD775820.1 | 1 | 113 |
More Information
Set Membership: |
Intragenic Exonic |