Assay Details
Target Gene Details
Entrez Gene ID: | 140559 |
Gene Name: | immunoglobulin superfamily, member 8 |
Gene Aliases: |
AA033172, AU041109, ESTM34, EWI-2, KCT-4, PGRL, R75384 |
Location: |
Chr.1:172312407-172319841 on Build GRCm38 |
Assay Gene Location: | Within Exon 4 |
Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
---|---|---|---|---|
Igsf8 | NM_080419.1 | 4 | 1094 | NP_536344.1 |
AF411055.1 | 4 | 965 | AAL02217.1 | |
AF439263.1 | 4 | 1077 | AAN63626.1 | |
AK140254.1 | 4 | 1107 | ||
AK146104.1 | 4 | 1107 | BAE26904.1 | |
AK146221.1 | 4 | 1113 | BAE26989.1 | |
AK150239.1 | 4 | 1113 | BAE29403.1 | |
AK158522.1 | 4 | 1115 | BAE34544.1 | |
AK168363.1 | 4 | 1113 | BAE40298.1 | |
AK179661.1 | ||||
AK191306.1 | ||||
AK207901.1 | ||||
AK213281.1 | ||||
BC019587.1 | 2 | 116 | AAH19587.1 | |
BC026464.1 | 3 | 900 | AAH26464.1 | |
BC048387.1 | 4 | 1094 | AAH48387.1 |
More Information
Set Membership: |
Intragenic Exonic |