Assay Details
Target Gene Details
Entrez Gene ID: | 383619 |
Gene Name: | absent in melanoma 2 |
Gene Aliases: |
Gm1313, Ifi210 |
Location: |
Chr.1:173349246-173466040 on Build GRCm38 |
Assay Gene Location: | Within Exon 5 |
Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
---|---|---|---|---|
Aim2 | NM_001013779.2 | 5 | 619 | NP_001013801.2 |
AK050724.1 | 5 | 619 | ||
DT921984.1 | 1 | 145 | ||
JQ894737.1 | 1 | 165 |
More Information
Set Membership: |
Intragenic Exonic |