Assay Details
Target Gene Details
Entrez Gene ID: | 18071 |
Gene Name: | nescient helix loop helix 1 |
Gene Aliases: |
Hen1, Nscl, Tal2, bHLHa35 |
Location: |
Chr.1:172052292-172057596 on Build GRCm38 |
Assay Gene Location: | Within Exon 1 |
Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
---|---|---|---|---|
Nhlh1 | NM_010916.2 | 1 | 211 | NP_035046.1 |
AK149315.1 | 1 | 212 | BAE28808.1 | |
BC051018.1 | 1 | 186 | AAH51018.1 | |
M82874.1 | 1 | 182 | AAA39840.1 |
More Information
Set Membership: |
Intragenic Exonic |