Assay Details
Target Gene Details
Entrez Gene ID: | 12477 |
Gene Name: | cytotoxic T-lymphocyte-associated protein 4 |
Gene Aliases: |
Cd152, Ctla-4, Ly-56 |
Location: |
Chr.1:60909025-60915832 on Build GRCm38 |
Assay Gene Location: | Within Exon 4 |
Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
---|---|---|---|---|
Ctla4 | NM_001281976.1 | 3 | 1010 | NP_001268905.1 |
NM_009843.4 | 4 | 1120 | NP_033973.2 | |
AK138891.1 | 4 | 1054 | ||
BC042741.1 | 4 | 1120 | AAH42741.1 | |
BC052683.1 | 4 | 1040 | AAH52683.1 | |
X05719.1 | 4 | 1031 | CAA29191.1 |
More Information
Set Membership: |
Intragenic Exonic |