Assay Details
Target Gene Details
Entrez Gene ID: | 226747 |
Gene Name: | AT hook containing transcription factor 1 |
Gene Aliases: |
6230412P20Rik, AV011447, Elys |
Location: |
Chr.1:179744894-179804015 on Build GRCm38 |
Assay Gene Location: | Within Exon 33 |
Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
---|---|---|---|---|
Ahctf1 | NM_026375.2 | 33 | 6112 | NP_080651.2 |
AB059278.1 | 33 | 5768 | BAB78517.1 | |
BC138343.1 | 33 | 5755 | ||
BC138344.1 | 33 | 5755 |
More Information
Set Membership: |
Intragenic Exonic |