Assay Details
Target Gene Details
Entrez Gene ID: | 16782 |
Gene Name: | laminin, gamma 2 |
Gene Aliases: |
AA589349 |
Location: |
Chr.1:153122756-153186447 on Build GRCm38 |
Assay Gene Location: | Within Exon 24 |
Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
---|---|---|---|---|
Lamc2 | NM_008485.3 | 24 | 3902 | NP_032511.3 |
DV658346.1 | 3 | 529 | ||
U43327.2 | 23 | 3629 | AAA85256.2 |
More Information
Set Membership: |
Intragenic Exonic |