Assay Details
Target Gene Details
Entrez Gene ID: | 383619 |
Gene Name: | absent in melanoma 2 |
Gene Aliases: |
Gm1313, Ifi210 |
Location: |
Chr.1:173349246-173466040 on Build GRCm38 |
Assay Gene Location: |
Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
---|---|---|---|---|
Aim2 |
Target Gene Details
Entrez Gene ID: | 94332 |
Gene Name: | cell adhesion molecule 3 |
Gene Aliases: |
BIgR, Igsf4b, Necl-1, Necl1, SynCAM3, Tsll1 |
Location: |
Chr.1:173334254-173367695 on Build GRCm38 |
Assay Gene Location: | Within Exon 1 |
Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
---|---|---|---|---|
Cadm3 | NM_053199.3 | 1 | 241 | NP_444429.1 |
AF195662.1 | 1 | 185 | AAG35584.1 | |
AK036352.1 | 1 | 195 | ||
AK038917.1 | 1 | 198 | BAC30168.1 | |
AK053077.1 | 1 | 195 | BAC35258.1 | |
AY059393.1 | AAL29691.1 | |||
BC115417.2 | 1 | 146 | ||
BC115418.2 | 1 | 146 | ||
BY289385.1 | 1 | 242 |