Assay Details
Target Gene Details
Entrez Gene ID: | 68691 |
Gene Name: | KAT8 regulatory NSL complex subunit 1-like |
Gene Aliases: |
1110028C15Rik, 1190030G24, AI427598, C430010P07Rik, mKIAA4189 |
Location: |
Chr.1:66719243-66817596 on Build GRCm38 |
Assay Gene Location: | Within Exon 2 |
Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
---|---|---|---|---|
Kansl1l | NM_001122738.1 | 2 | 600 | NP_001116210.1 |
NM_177645.4 | 2 | 600 | NP_808313.3 | |
AK081413.1 | 3 | 950 | BAC38212.1 | |
AK138707.1 | 1 | 431 | ||
AK145440.1 | 2 | 571 | ||
AK170981.1 | 2 | 629 | BAE42157.1 | |
AK172645.1 | 2 | 552 | ||
AK220534.1 | 2 | 551 | BAD90312.1 | |
BC141036.1 | 3 | 510 |