Assay Details
Target Gene Details
Entrez Gene ID: | 12227 |
Gene Name: | B cell translocation gene 2, anti-proliferative |
Gene Aliases: |
AA959598, APRO1, Pc3, TIS21 |
Location: |
Chr.1:134074865-134079155 on Build GRCm38 |
Assay Gene Location: | Within Exon 2 |
Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
---|---|---|---|---|
Btg2 | NM_007570.2 | 2 | 2138 | NP_031596.1 |
AK133796.1 | 2 | 2103 | BAE21847.1 | |
AK169269.1 | 2 | 2103 | BAE41030.1 | |
AK169710.1 | 1 | 1437 | ||
AK171400.1 | 1 | 793 | ||
BC132259.1 | 2 | 2088 | ||
BC138639.1 | 2 | 2088 |