Assay Details
Target Gene Details
Entrez Gene ID: | 246229 |
Gene Name: | basic, immunoglobulin-like variable motif containing |
Gene Aliases: |
AI854267 |
Location: |
Chr.1:44118888-44144771 on Build GRCm38 |
Assay Gene Location: | Within Exon 10 |
Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
---|---|---|---|---|
Bivm | NM_144558.4 | 10 | 3240 | NP_653141.3 |
AF411386.1 | 11 | 3243 | AAM45762.1 | |
AK032375.1 | 10 | 3200 | BAC27843.1 | |
AK034329.1 | 10 | 3202 | BAC28676.1 | |
AK088931.1 | 1 | 1253 | ||
BC082584.1 | 10 | 3046 | AAH82584.1 | |
CJ309003.1 | 1 | 116 |
More Information
Set Membership: |
Intragenic Exonic |