Assay Details
Target Gene Details
Entrez Gene ID: | 16865 |
Gene Name: | eukaryotic translation initiation factor 2D |
Gene Aliases: |
D1Ertd5e, Lgtn |
Location: |
Chr.1:131153170-131187659 on Build GRCm38 |
Assay Gene Location: |
Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
---|---|---|---|---|
Eif2d | AK036154.1 | BAC29323.1 |
Target Gene Details
Entrez Gene ID: | 54354 |
Gene Name: | Ras association (RalGDS/AF-6) domain family member 5 |
Gene Aliases: |
1300019G20Rik, AU042887, Maxp1, Nore1, Nore1A, Nore1B, Rapl |
Location: |
Chr.1:131176410-131245178 on Build GRCm38 |
Assay Gene Location: | Within Exon 4 |
Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
---|---|---|---|---|
Rassf5 | NM_018750.3 | 4 | 981 | NP_061220.2 |
AF053959.1 | 4 | 963 | AAC08580.1 | |
AK005059.2 | 4 | 977 | ||
AK029321.1 | ||||
AK088751.1 | 3 | 616 | BAC40546.1 | |
AK131147.1 | 3 | 574 | BAD21397.1 | |
AK155534.1 | 4 | 977 | BAE33312.1 | |
AK155869.1 | 3 | 653 | BAE33472.1 | |
AY261333.1 | 3 | 592 | AAP83361.1 | |
BC089605.1 | 4 | 983 | AAH89605.1 |
More Information
Set Membership: |
Intragenic Exonic Non-exonic |