Assay Details
Target Gene Details
Entrez Gene ID: | 11363 |
Gene Name: | acyl-Coenzyme A dehydrogenase, long-chain |
Gene Aliases: |
AA960361, AU018452, C79855, LCAD |
Location: |
Chr.1:66830839-66863309 on Build GRCm38 |
Assay Gene Location: | Within Exon 11 |
Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
---|---|---|---|---|
Acadl | NM_007381.4 | 11 | 1372 | NP_031407.2 |
AK005140.1 | 11 | 1331 | BAB23838.1 | |
AK018319.1 | 10 | 1148 | BAB31161.1 | |
AK151845.1 | 11 | 1341 | ||
AK167537.1 | 11 | 1339 | ||
BC027412.1 | 11 | 1329 | AAH27412.1 | |
U21489.1 | 11 | 1312 | AAC52329.1 |
More Information
Set Membership: |
Intragenic Exonic |