Assay Details
Target Gene Details
Entrez Gene ID: | 269132 |
Gene Name: | collagen beta(1-O)galactosyltransferase 2 |
Gene Aliases: |
AI427652, D730024P12, Glt25d2 |
Location: |
Chr.1:152399867-152510695 on Build GRCm38 |
Assay Gene Location: | Within Exon 1 |
Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
---|---|---|---|---|
Colgalt2 | NM_177756.4 | 1 | 267 | NP_808424.3 |
AK052838.1 | 1 | 268 | BAC35169.1 | |
AK076222.1 | ||||
AK085716.1 | 1 | 305 | ||
AK134084.1 | 1 | 233 | ||
BC068118.1 | 1 | 213 | AAH68118.1 |
More Information
Set Membership: |
Intragenic Exonic |