Assay Details
Target Gene Details
Entrez Gene ID: | 320742 |
Gene Name: | RIKEN cDNA A230072C01 gene |
Gene Aliases: |
- |
Location: |
Chr.X:20961676-20987349 on Build GRCm38 |
Assay Gene Location: | Overlaps - Exon 1 |
Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
---|---|---|---|---|
A230072C01Rik | NR_027445.1 | |||
AK029359.1 | ||||
BY019647.1 |
Target Gene Details
Entrez Gene ID: | 22294 |
Gene Name: | ubiquitously expressed transcript |
Gene Aliases: |
0910002B17Rik |
Location: |
Chr.X:20951665-20962035 on Build GRCm38 |
Assay Gene Location: | Overlaps Intron 2 - Exon 2 |
Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
---|---|---|---|---|
Uxt | NM_013840.3 | NP_038868.2 | ||
AF092738.1 | AAD28699.1 | |||
AK027917.1 | BAC25665.1 | |||
AK086822.1 | ||||
AK163983.1 | BAE37569.1 | |||
BC029258.1 | AAH29258.1 | |||
BC082294.1 | AAH82294.1 |
More Information
Set Membership: |
Intragenic Non-exonic |