Assay Details
Target Gene Details
Entrez Gene ID: | 209837 |
Gene Name: | solute carrier family 38, member 5 |
Gene Aliases: |
C81234, E330031E14, JM24, SN2 |
Location: |
Chr.X:8271381-8280179 on Build GRCm38 |
Assay Gene Location: | Overlaps Exon 11 - Intron 11 |
Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
---|---|---|---|---|
Slc38a5 | NM_172479.3 | NP_766067.2 | ||
AK054485.1 | BAC35799.1 | |||
AK077667.1 | BAC36941.1 | |||
AK155928.1 | BAE33507.1 | |||
AK158125.1 | BAE34371.1 | |||
BC058856.1 | AAH58856.1 | |||
BC152401.1 |
More Information
Set Membership: |
Intragenic Non-exonic |