Assay Details
Target Gene Details
Entrez Gene ID: | 194744 |
Gene Name: | solute carrier family 25, member 43 |
Gene Aliases: |
Gm8 |
Location: |
Chr.X:36743549-36777307 on Build GRCm38 |
Assay Gene Location: | Within Exon 2 |
Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
---|---|---|---|---|
Slc25a43 | NM_001085497.2 | 2 | 429 | NP_001078966.1 |
NR_075096.1 | 2 | 429 | ||
NR_075097.1 | ||||
AK153028.1 | ||||
BC147709.1 | 2 | 429 | ||
BC147717.1 | 2 | 429 |
More Information
Set Membership: |
Intragenic Exonic |