Assay Details
Target Gene Details
Entrez Gene ID: | 20523 |
Gene Name: | solute carrier family 25 (mitochondrial carrier, brain), member 14 |
Gene Aliases: |
BMCP1, UCP5, UCP5L, UCP5S |
Location: |
Chr.X:48623418-48662298 on Build GRCm38 |
Assay Gene Location: | Within Exon 2 |
Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
---|---|---|---|---|
Slc25a14 | NM_001166450.2 | 2 | 272 | NP_001159922.1 |
NM_001290703.1 | 2 | 368 | NP_001277632.1 | |
NM_001290704.1 | 2 | 368 | NP_001277633.1 | |
NM_001290705.1 | NP_001277634.1 | |||
NM_011398.3 | 2 | 368 | NP_035528.1 | |
AF076981.1 | 2 | 368 | AAD03674.1 | |
AF535150.1 | 2 | 368 | AAN63885.1 | |
AF535151.1 | 2 | 368 | AAN63886.1 | |
AK051145.1 | 1 | 320 | ||
AK148277.1 | 1 | 320 | ||
BC048692.1 | 1 | 82 | AAH48692.1 | |
CX224035.1 | 1 | 117 | ||
CX238926.1 | 2 | 133 | ||
CX244278.1 | 2 | 366 |
More Information
Set Membership: |
Intragenic Exonic |