Assay Details
Target Gene Details
Entrez Gene ID: | 18846 |
Gene Name: | plexin A3 |
Gene Aliases: |
PlexA3, Plxa3, Plxn3, Plxn4, SEX, mKIAA4078 |
Location: |
Chr.X:74329066-74344689 on Build GRCm38 |
Assay Gene Location: | Within Exon 9 |
Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
---|---|---|---|---|
Plxna3 | NM_008883.2 | 9 | 2057 | NP_032909.2 |
AK220285.1 | 7 | 782 | BAD90210.1 | |
BC093482.1 | 9 | 2040 | ||
D86950.1 | 9 | 2023 | BAA13190.1 |