Assay Details
Target Gene Details
Entrez Gene ID: | 18846 |
Gene Name: | plexin A3 |
Gene Aliases: |
PlexA3, Plxa3, Plxn3, Plxn4, SEX, mKIAA4078 |
Location: |
Chr.X:74329066-74344689 on Build GRCm38 |
Assay Gene Location: | Overlaps Exon 25 - Intron 25 |
Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
---|---|---|---|---|
Plxna3 | NM_008883.2 | NP_032909.2 | ||
AJ748653.1 | CAG38687.1 | |||
AK049319.1 | BAC33681.1 | |||
AK220285.1 | BAD90210.1 | |||
BC093482.1 | ||||
D86950.1 | BAA13190.1 |