Assay Details
Target Gene Details
Entrez Gene ID: | 209268 |
Gene Name: | immunoglobulin superfamily, member 1 |
Gene Aliases: |
5330413N23, 5530402E03, AI747649, InhBP/p120, mKIAA0364 |
Location: |
Chr.X:49782536-49797906 on Build GRCm38 |
Assay Gene Location: | Within Exon 14 |
Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
---|---|---|---|---|
Igsf1 | NM_177591.4 | NP_808259.2 | ||
NM_183335.2 | 13 | 2378 | NP_899178.2 | |
NM_183336.2 | NP_899179.1 | |||
AK030701.1 | ||||
AK076427.1 | BAC36338.1 | |||
AK129126.1 | BAC97936.1 | |||
AY227771.1 | AAP57079.1 | |||
AY227773.1 | 13 | 2378 | AAP57081.1 | |
AY227774.1 | AAP57082.1 | |||
BC119264.1 | ||||
BC119266.1 |
More Information
Set Membership: |
Intragenic Exonic |