Assay Details
Target Gene Details
Entrez Gene ID: | 236727 |
Gene Name: | solute carrier family 9 (sodium/hydrogen exchanger), member 7 |
Gene Aliases: |
A530087D17Rik, NHE7 |
Location: |
Chr.X:20098851-20291970 on Build GRCm38 |
Assay Gene Location: | Overlaps Intron 9 - Exon 10 |
Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
---|---|---|---|---|
Slc9a7 | NM_177353.3 | NP_796327.1 | ||
AK041169.1 | BAC30848.1 | |||
AK043187.1 | ||||
BC058750.1 | AAH58750.1 |
More Information
Set Membership: |
Intragenic Non-exonic |