Assay Details
Target Gene Details
Entrez Gene ID: | 236794 |
Gene Name: | solute carrier family 9 (sodium/hydrogen exchanger), member 6 |
Gene Aliases: |
3732426M05, 6430520C02Rik, NHE6, mKIAA0267 |
Location: |
Chr.X:56609557-56664230 on Build GRCm38 |
Assay Gene Location: | Within Exon 12 |
Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
---|---|---|---|---|
Slc9a6 | NM_172780.3 | 12 | 1559 | NP_766368.2 |
AK032326.1 | 12 | 1540 | BAC27816.1 | |
AK122232.1 | 12 | 1530 | BAC65514.1 | |
BC130221.1 | 12 | 1544 |
More Information
Set Membership: |
Intragenic Exonic |