Assay Details
Target Gene Details
Entrez Gene ID: | 18823 |
Gene Name: | proteolipid protein (myelin) 1 |
Gene Aliases: |
DM20, Plp, jimpy, jp, msd, rsh |
Location: |
Chr.X:136822746-136838582 on Build GRCm38 |
Assay Gene Location: | Within Exon 2 |
Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
---|---|---|---|---|
Plp1 | NM_001290561.1 | 2 | 265 | NP_001277490.1 |
NM_001290562.1 | 2 | 265 | NP_001277491.1 | |
NM_011123.3 | 2 | 265 | NP_035253.1 | |
AK077564.1 | 2 | 221 | ||
AK134554.1 | 2 | 256 | BAE22184.1 | |
AK160872.1 | 2 | 260 | BAE36061.1 | |
AK192772.1 | 2 | 148 | ||
AK203421.1 | 2 | 148 | ||
BC027010.1 | 2 | 175 | AAH27010.1 | |
BY282535.1 | 2 | 265 | ||
M14674.1 | 1 | 99 | AAA39955.1 | |
M15442.1 | 2 | 244 | AAA39951.1 | |
M16472.1 | 1 | 99 | AAA39950.1 | |
M20752.1 | 1 | 99 | AAA39956.1 |
More Information
Set Membership: |
Intragenic Exonic |