Assay Details
Target Gene Details
Entrez Gene ID: | 14069 |
Gene Name: | coagulation factor VIII |
Gene Aliases: |
Cf-8, Cf8, FVIII |
Location: |
Chr.X:75172715-75383525 on Build GRCm38 |
Assay Gene Location: | Within Exon 11 |
Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
---|---|---|---|---|
F8 | NM_001161373.1 | 11 | 2102 | NP_001154845.1 |
NM_001161374.1 | 11 | 2102 | NP_001154846.1 | |
NM_007977.2 | 11 | 2102 | NP_032003.2 | |
BC138347.1 | 11 | 1983 | ||
BC145245.1 | 11 | 1983 | ||
L05573.1 | 11 | 2100 | AAA37385.1 |
More Information
Set Membership: |
Intragenic Exonic |