Assay Details
Target Gene Details
Entrez Gene ID: | 210710 |
Gene Name: | growth factor receptor bound protein 2-associated protein 3 |
Gene Aliases: |
5930433H21Rik |
Location: |
Chr.X:74988545-75084905 on Build GRCm38 |
Assay Gene Location: | Overlaps Intron 7 - Exon 7 |
Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
---|---|---|---|---|
Gab3 | NM_181584.4 | NP_853615.1 | ||
AJ748654.1 | CAG38688.1 | |||
AK031227.1 | BAC27308.1 | |||
AY057988.1 | AAL25824.1 | |||
BB634123.1 | ||||
BC141240.1 |
More Information
Set Membership: |
Intragenic Non-exonic |