Assay Details
Target Gene Details
Entrez Gene ID: | 209224 |
Gene Name: | ecto-NOX disulfide-thiol exchanger 2 |
Gene Aliases: |
APK1, Cova1, tNOX |
Location: |
Chr.X:49009707-49288245 on Build GRCm38 |
Assay Gene Location: | Within Exon 1 |
Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
---|---|---|---|---|
Enox2 | NM_001271447.1 | 1 | 90 | NP_001258376.1 |
NM_001271448.1 | 1 | 90 | NP_001258377.1 | |
NM_001271449.1 | 1 | 90 | NP_001258378.1 | |
NM_001271450.1 | 1 | 90 | NP_001258379.1 | |
NM_001271451.1 | 1 | 90 | NP_001258380.1 | |
NM_145951.5 | NP_666063.1 | |||
AK028343.1 | 1 | 91 | ||
AK034058.1 | BAC28565.1 | |||
AK041822.1 | 1 | 91 | BAC31077.1 | |
AK077508.1 | ||||
AK133956.1 | BAE21951.1 | |||
AK137736.1 | BAE23482.1 | |||
AK139897.1 | BAE24174.1 | |||
BC025915.1 | AAH25915.1 |
More Information
Set Membership: |
Intragenic Exonic |