Assay Details
Target Gene Details
Entrez Gene ID: | 245536 |
Gene Name: | predicted gene 614 |
Gene Aliases: |
Il2rg |
Location: |
Chr.X:101261377-101263992 on Build GRCm38 |
Assay Gene Location: | Overlaps Intron 4 - Exon 5 |
Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
---|---|---|---|---|
Gm614 | NM_001033362.2 | NP_001028534.1 | ||
AK131750.1 | BAE20796.1 | |||
BC147523.1 | ||||
BC147537.1 |