Assay Details
Target Gene Details
Entrez Gene ID: | 14168 |
Gene Name: | fibroblast growth factor 13 |
Gene Aliases: |
Fhf2 |
Location: |
Chr.X:59062139-59585572 on Build GRCm38 |
Assay Gene Location: | Overlaps Intron 6 - Exon 6 |
Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
---|---|---|---|---|
Fgf13 | NM_001290414.1 | NP_001277343.1 | ||
NM_001290415.1 | NP_001277344.1 | |||
NM_010200.3 | NP_034330.2 | |||
AF020737.1 | AAB71606.1 | |||
AF199608.1 | AAF31395.1 | |||
AF199614.1 | AAF31401.1 | |||
AF199615.1 | ||||
AK141848.1 | BAE24857.1 | |||
BC018238.1 | AAH18238.1 | |||
CJ064772.1 | ||||
U66202.1 | AAB18918.1 |
More Information
Set Membership: |
Intragenic Non-exonic |