Assay Details
Target Gene Details
Entrez Gene ID: | 56292 |
Gene Name: | N(alpha)-acetyltransferase 10, NatA catalytic subunit |
Gene Aliases: |
2310039H09Rik, Ard1, Ard1a, Te2 |
Location: |
Chr.X:73916870-73921944 on Build GRCm38 |
Assay Gene Location: | Overlaps - Exon 1 |
Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
---|---|---|---|---|
Naa10 | NM_001177965.1 | NP_001171436.1 | ||
NM_019870.3 | NP_063923.1 | |||
AK003806.1 | BAE43151.1 | |||
AK009697.1 | BAC25266.1 | |||
AK014469.1 | BAB29373.1 | |||
AK078700.1 | BAC37364.1 | |||
AY183136.1 | AAO66339.1 | |||
AY183137.1 | ||||
BY101790.1 |
More Information
Set Membership: |
Intragenic Non-exonic |