Assay Details
Target Gene Details
Entrez Gene ID: | 109113 |
Gene Name: | ubiquitin-like, containing PHD and RING finger domains 2 |
Gene Aliases: |
2310065A22Rik, AI426270, AW214556, D130071B19Rik, Nirf |
Location: |
Chr.19:30030433-30093729 on Build GRCm38 |
Assay Gene Location: | Overlaps Intron 13 - Exon 14 |
Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
---|---|---|---|---|
Uhrf2 | NM_144873.2 | NP_659122.2 | ||
AB116653.1 | BAC81739.1 | |||
AF274047.1 | AAM33798.1 | |||
AK075912.1 | ||||
AK170648.1 | BAE41935.1 | |||
BC060241.1 | AAH60241.1 |
More Information
Set Membership: |
Intragenic Non-exonic |