Assay Details
Target Gene Details
Entrez Gene ID: | 226026 |
Gene Name: | structural maintenance of chromosomes 5 |
Gene Aliases: |
MLZ-453, SMC-5, Smc5l1, mKIAA0594, mSMC5 |
Location: |
Chr.19:23206441-23273908 on Build GRCm38 |
Assay Gene Location: | Within Exon 24 |
Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
---|---|---|---|---|
Smc5 | NM_001252684.1 | 23 | 3063 | NP_001239613.1 |
NM_001252685.1 | 22 | 2850 | NP_001239614.1 | |
NM_153808.2 | 22 | 3021 | NP_722503.1 | |
AJ534941.1 | 23 | 2984 | CAD59184.1 | |
AK122321.1 | 15 | 5214 | BAC65603.3 | |
AK161254.1 | 22 | 2853 | ||
BC007174.1 | 14 | 1619 | AAH07174.1 | |
BC038345.1 | 22 | 3001 | AAH38345.1 |