Assay Details
Target Gene Details
Entrez Gene ID: | 225887 |
Gene Name: | NADH dehydrogenase (ubiquinone) Fe-S protein 8 |
Gene Aliases: |
BC021616, CI-23kD, TYKY |
Location: |
Chr.19:3908863-3912774 on Build GRCm38 |
Assay Gene Location: | Within Exon 7 |
Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
---|---|---|---|---|
Ndufs8 | NM_001271443.1 | 7 | 682 | NP_001258372.1 |
NM_001271444.1 | 7 | 922 | NP_001258373.1 | |
NM_144870.5 | 7 | 925 | NP_659119.2 | |
AK135069.1 | 7 | 683 | BAE22408.1 | |
AK149674.1 | 7 | 883 | BAE29018.1 | |
AK153182.1 | 7 | 869 | BAE31784.1 | |
AK153900.1 | 7 | 659 | BAE32243.1 | |
AK164901.1 | 7 | 659 | BAE37960.1 | |
AK167907.1 | 7 | 660 | BAE39914.1 | |
BC021616.1 | 7 | 643 | AAH21616.2 | |
BC086766.1 | 7 | 636 | AAH86766.1 | |
BP775598.1 | 1 | 158 |
More Information
Set Membership: |
Intragenic Exonic |