Assay Details
Target Gene Details
Entrez Gene ID: | 18938 |
Gene Name: | protein phosphatase 1, regulatory (inhibitor) subunit 14B |
Gene Aliases: |
AOM172, PHI-1, PLCB3N, Png |
Location: |
Chr.19:6975048-6977324 on Build GRCm38 |
Assay Gene Location: | Overlaps Exon 2 - Intron 2 |
Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
---|---|---|---|---|
Ppp1r14b | NM_008889.2 | NP_032915.2 | ||
AK141152.1 | ||||
AK165792.1 | BAE38382.1 | |||
BC082545.1 | AAH82545.2 | |||
BC125650.1 | ||||
BC125654.1 | ||||
CF739461.1 | ||||
X97490.1 | CAA66121.1 |
More Information
Set Membership: |
Intragenic Non-exonic |