Assay Details
Target Gene Details
Entrez Gene ID: | 66990 |
Gene Name: | transmembrane protein 134 |
Gene Aliases: |
2410001H17Rik, AI463452 |
Location: |
Chr.19:4125960-4132307 on Build GRCm38 |
Assay Gene Location: | Overlaps Intron 3 - Exon 4 |
Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
---|---|---|---|---|
Tmem134 | NM_001078649.1 | NP_001072117.1 | ||
NM_025889.2 | NP_080165.1 | |||
AI789565.1 | ||||
AK002220.1 | BAB21944.1 | |||
AK008515.1 | BAB25713.1 | |||
AK009263.1 | 3 | 506 | ||
AK009745.1 | BAB26475.1 | |||
AK010323.1 | BAC25290.1 | |||
AK129014.1 | BAC87670.1 | |||
AK166519.1 | BAE38823.1 | |||
AK178280.1 | ||||
AK183549.1 | ||||
AK195169.1 | ||||
BC026765.1 | AAH26765.1 | |||
CF983315.1 |