Assay Details
Target Gene Details
Entrez Gene ID: | 67155 |
Gene Name: | SWI/SNF related, matrix associated, actin dependent regulator of chromatin, subfamily a, member 2 |
Gene Aliases: |
2610209L14Rik, SNF2alpha, Snf2l2, brahma, brm |
Location: |
Chr.19:26605089-26778321 on Build GRCm38 |
Assay Gene Location: | Within Exon 19 |
Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
---|---|---|---|---|
Smarca2 | NM_011416.2 | 19 | 2967 | NP_035546.2 |
AK134308.1 | 5 | 552 | ||
AK135871.1 | 3 | 318 | BAE22708.1 | |
AK147322.1 | 19 | 3168 | BAE27845.1 | |
BC056199.1 | 9 | 1065 | AAH56199.1 | |
BC075641.1 | 19 | 2949 | AAH75641.1 |
More Information
Set Membership: |
Intragenic Exonic |