Assay Details
Target Gene Details
Entrez Gene ID: | 18563 |
Gene Name: | pyruvate carboxylase |
Gene Aliases: |
Pc, Pcb |
Location: |
Chr.19:4510472-4621752 on Build GRCm38 |
Assay Gene Location: | Within Exon 11 |
Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
---|---|---|---|---|
Pcx | NM_001162946.1 | 10 | 1214 | NP_001156418.1 |
NM_008797.3 | 9 | 1191 | NP_032823.2 | |
AK077665.1 | BAC36939.1 | |||
AK148328.1 | 10 | 1161 | BAE28484.1 | |
AK170596.1 | 10 | 1174 | BAE41902.1 | |
AK170871.1 | 10 | 1164 | ||
AK172320.1 | 10 | 1142 | BAE42943.1 | |
BC055030.1 | 10 | 1149 | AAH55030.1 | |
L09192.1 | 9 | 1167 | AAA39737.1 | |
M97957.1 | 4 | 339 | AAA39897.1 |
More Information
Set Membership: |
Intragenic Exonic |