Assay Details
Target Gene Details
Entrez Gene ID: | 18563 |
Gene Name: | pyruvate carboxylase |
Gene Aliases: |
Pc, Pcb |
Location: |
Chr.19:4510472-4621752 on Build GRCm38 |
Assay Gene Location: | Overlaps Exon 8 - Intron 8 |
Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
---|---|---|---|---|
Pcx | NM_001162946.1 | NP_001156418.1 | ||
NM_008797.3 | NP_032823.2 | |||
AK148328.1 | BAE28484.1 | |||
AK170596.1 | BAE41902.1 | |||
AK170871.1 | ||||
AK172320.1 | BAE42943.1 | |||
BC055030.1 | AAH55030.1 | |||
L09192.1 | AAA39737.1 | |||
M97957.1 | AAA39897.1 |
More Information
Set Membership: |
Intragenic Non-exonic |