Assay Details
Target Gene Details
Entrez Gene ID: | 19726 |
Gene Name: | regulatory factor X, 3 (influences HLA class II expression) |
Gene Aliases: |
C230093O12Rik, MRFX3 |
Location: |
Chr.19:27761718-28011698 on Build GRCm38 |
Assay Gene Location: | Overlaps Intron 4 - Exon 5 |
Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
---|---|---|---|---|
Rfx3 | NM_001166414.1 | NP_001159886.1 | ||
NM_011265.3 | NP_035395.2 | |||
AK030008.1 | BAC26730.1 | |||
AK041120.1 | BAC30829.1 | |||
AK140928.1 | ||||
BC017598.1 | AAH17598.1 |
More Information
Set Membership: |
Intragenic Non-exonic |