Assay Details
Target Gene Details
Entrez Gene ID: | 107328 |
Gene Name: | tRNA phosphotransferase 1 |
Gene Aliases: |
AU079016, AW045591, Tpt1, Tpt1h |
Location: |
Chr.19:6996131-6999048 on Build GRCm38 |
Assay Gene Location: | Overlaps Exon 4 - Intron 4 |
Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
---|---|---|---|---|
Trpt1 | NM_153597.2 | NP_705825.2 | ||
NR_027351.1 | ||||
AK038830.1 | ||||
AK090183.1 | ||||
BC027575.1 | AAH27575.1 | |||
DQ113648.1 |
More Information
Set Membership: |
Intragenic Non-exonic |