Assay Details
Target Gene Details
Entrez Gene ID: | 13041 |
Gene Name: | cathepsin W |
Gene Aliases: |
lymphopain |
Location: |
Chr.19:5465240-5470387 on Build GRCm38 |
Assay Gene Location: | Overlaps Intron 7 - Exon 7 |
Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
---|---|---|---|---|
Ctsw | NM_009985.4 | NP_034115.2 | ||
AF014941.1 | AAB82455.1 | |||
AI450305.1 | ||||
AK088375.1 | BAC40314.1 | |||
AK163626.1 | ||||
BE447240.1 |
More Information
Set Membership: |
Intragenic Non-exonic |