Assay Details
Target Gene Details
Entrez Gene ID: | 16952 |
Gene Name: | annexin A1 |
Gene Aliases: |
Anx-1, Anx-A1, C430014K04Rik, Lpc-1, Lpc1 |
Location: |
Chr.19:20373434-20390671 on Build GRCm38 |
Assay Gene Location: | Overlaps Intron 5 - Exon 6 |
Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
---|---|---|---|---|
Anxa1 | NM_010730.2 | NP_034860.2 | ||
AK002681.1 | ||||
AK140838.1 | BAE24494.1 | |||
AK150564.1 | BAE29662.1 | |||
AK150718.1 | BAE29796.1 | |||
AK153156.1 | BAE31766.1 | |||
AK153490.1 | BAE32038.1 | |||
AK167612.1 | BAE39665.1 | |||
BC002289.1 | AAH02289.1 | |||
BC004594.1 | AAH04594.1 | |||
CT009771.1 | CAJ18120.1 | |||
CT010298.1 | CAJ18506.1 | |||
EU684130.1 | ||||
M24554.1 | AAA39420.1 | |||
X07486.1 | CAA30371.1 |
More Information
Set Membership: |
Intragenic Non-exonic |