Assay Details
Target Gene Details
Entrez Gene ID: | 81601 |
Gene Name: | K(lysine) acetyltransferase 5 |
Gene Aliases: |
AI839539, CPLA2, Htatip, Htatip1, PLIP, Tip55, Tip60 |
Location: |
Chr.19:5603014-5610232 on Build GRCm38 |
Assay Gene Location: | Within Exon 13 |
Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
---|---|---|---|---|
Kat5 | NM_001199247.1 | 13 | 1389 | NP_001186176.1 |
NM_001199248.1 | 12 | 1488 | NP_001186177.1 | |
NM_178637.2 | 14 | 1545 | NP_848752.1 | |
NR_037603.1 | 11 | 1421 | ||
AB055409.1 | 11 | 1399 | BAC53807.1 | |
AF528194.1 | 10 | 1099 | AAN77140.1 | |
AF528195.1 | 13 | 1480 | AAN77141.1 | |
AF528196.1 | 12 | 1324 | AAN77142.1 | |
AK133843.1 | 2 | 135 | ||
AK146352.1 | 14 | 1502 | BAE27104.1 | |
AK159568.1 | 12 | 1421 | ||
AK160077.1 | 12 | 1421 | ||
AK177994.1 | ||||
AK187951.1 | ||||
BC110675.1 | 15 | 1779 | AAI10676.1 | |
BC129967.1 | 12 | 1323 | ||
BC129968.1 | 12 | 1324 | ||
DQ139980.1 | 14 | 1773 | AAZ67923.1 | |
DV648619.1 | 1 | 55 |
More Information
Set Membership: |
Intragenic Exonic |