Assay Details
Target Gene Details
Entrez Gene ID: | 107765 |
Gene Name: | ankyrin repeat domain 1 (cardiac muscle) |
Gene Aliases: |
Alrp, CARP, Crap, MARP1 |
Location: |
Chr.19:36111965-36119844 on Build GRCm38 |
Assay Gene Location: | Overlaps Intron 1 - Exon 1 |
Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
---|---|---|---|---|
Ankrd1 | NM_013468.3 | NP_038496.2 | ||
AF041847.1 | AAC03533.1 | |||
AK009655.1 | BAB26419.1 | |||
AK009959.1 | BAB26611.1 | |||
AK145944.1 | BAE26773.1 | |||
AK146471.1 | BAE27197.1 | |||
AK146627.1 | BAE27316.1 | |||
AK146940.1 | BAE27549.1 | |||
BC037138.1 | AAH37138.1 |